Magnesium sulfate heptahydrate 99+%
Supplier: Thermo Scientific Chemicals
A14491-0B
A14491-0I
CAAAAA14491-0BEA
78.05
CAD
CAAAAA14491-0B
CAAAAA14491-0I
Magnesium sulfate heptahydrate 99+%
Magnesium sulphate heptahydrate
Purity: 99+%
CAS No.: 10034-99-8
Molecular Formula: MgSO4.7H2O
Formula Weight: 246.48 (120.37anhy)
Odor: Odorless
Merck Reference: 14,5691
MDL No.: MFCD00149785
Formula:
MgSO₄·7H₂O Melting Pt: 1124 °C Density: 1.68 Storage Temperature: Ambient |
MDL Number:
MFCD00149785 CAS Number: 10034-99-8 EINECS: 231-298-2 Merck Index: 14,5691 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...