Nickel(II) chloride hexahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickel dichloride hexahydrate

A14366-0B A14366-0I A14366-30
CAAAAA14366-0BEA 244.9 CAD
CAAAAA14366-0B CAAAAA14366-0I CAAAAA14366-30
Nickel(II) chloride hexahydrate 98%
Nickel (II) chloride hexahydrate

Purity: 98% CAS No.: 7791-20-0 Molecular Formula: Cl2Ni.6H2O Formula Weight: 237.71 (129.60anhy) Odor: Odorless pH: 4.9 Melting Point: 140° -H2O Merck Reference: 14,6505 UN No.: 3288 MDL No.: MFCD00149809

Formula: NiCl₂·6H₂O
MW: 237.69 g/mol
Melting Pt: 140 °C (-H₂O) (–H₂O of crystall.)
Density: 1.92
Storage Temperature: Ambient
MDL Number: MFCD00149809
CAS Number: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1 III
Merck Index: 14,6505

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR