Eugenol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole

A14332-22 A14332-36
CAAAAA14332-22EA 70.94 CAD
CAAAAA14332-22 CAAAAA14332-36
Eugenol 99%
Eugenol

Internal standard in GC assays for presence of thymol in biological samples


Purity: 99% CAS No.: 97-53-0 Molecular Formula: C10H12O2 Formula Weight: 164.21 Flash Point: 127°(260°F) Refractive Index: 1.5410 Odor: Pungent Melting Point: -10° Boiling Point: 252-253° Beilstein Reference: 1366759 Merck Reference: 14,3898 MDL No.: MFCD00008654

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 252…253 °C
Melting Pt: –10 °C
Density: 1.068
Flash Pt: 127 °C(260 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008654
CAS Number: 97-53-0
EINECS: 202-589-1
Merck Index: 14,3898

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR