Polyvinylpyrrolidone, M.W. ∼ 58,000 (K29-32)

Supplier: Thermo Scientific Chemicals

A14315-0E A14315-22 A14315-36
CAAAAA14315-0EEA 694.55 CAD
CAAAAA14315-0E CAAAAA14315-22 CAAAAA14315-36
Polyvinylpyrrolidone, M.W. ∼ 58,000 (K29-32)
Polyvinylpyrrolidone

A cryoprotectant, also useful in plant cell culture research


CAS No.: 9003-39-8 Molecular Formula: (C6H9NO)n Merck Reference: 14,7697 MDL No.: MFCD00149016

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8
Merck Index: 14,7697

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR