Magnesium hydrogen orthophosphate trihydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Magnesium hydrogen phosphate trihydrate, Magnesium phosphate dibasic trihydrate

A14279-0B A14279-30
CAAAAA14279-0BEA 106.97 CAD
CAAAAA14279-0B CAAAAA14279-30
Magnesium hydrogen orthophosphate trihydrate 99%
Magnesium hydrogen orthophosphate trihydrate

Purity: 99% CAS No.: 7782-75-4 Molecular Formula: MgHPO4.3H2O Formula Weight: 174.34 (120.28anhy) Odor: Odorless Merck Reference: 14,5681 MDL No.: MFCD00150390

Formula: HMgO₄P·3H₂O
Density: 2.13
Storage Temperature: Ambient
MDL Number: MFCD00150390
CAS Number: 7782-75-4
EINECS: 231-823-5
Merck Index: 14,5681

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR