Dimethyl methylphosphonate 97%

Supplier: Thermo Scientific Chemicals
Danger

A14268-09 A14268-36 A14268-22
CAAAAA14268-09EA 51.19 CAD
CAAAAA14268-09 CAAAAA14268-36 CAAAA14268-22
Dimethyl methylphosphonate 97%
Dimethyl methylphosphonate

Purity: 97% CAS No.: 756-79-6 Molecular Formula: C3H9O3P Formula Weight: 124.08 Flash Point: 68°(154°F) Refractive Index: 1.4130 Boiling Point: 180-181° Beilstein Reference: 878263 Merck Reference: 14,3391 MDL No.: MFCD00008349

Formula: C₃H₉O₃P
Boiling Pt: 180…181 °C
Density: 1.16
Flash Pt: 68 °C(154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008349
CAS Number: 756-79-6
EINECS: 212-052-3
Merck Index: 14,3391

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR