1H-Pyrazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyrazole, 1H-Pyrazole

A14186-14 A14186-22
CAAAAA14186-14EA 91.8 CAD
CAAAAA14186-14 CAAAAA14186-22
1H-Pyrazole 98%
1H-Pyrazole

Purity: 98% CAS No.: 288-13-1 Molecular Formula: C3H4N2 Formula Weight: 68.08 Odor: Unpleasant Melting Point: 66-70° Boiling Point: 186-188° Beilstein Reference: 103775 Merck Reference: 14,7960 MDL No.: MFCD00005234

Formula: C₃H₄N₂
Boiling Pt: 186…188 °C
Melting Pt: 66…70 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005234
CAS Number: 288-13-1
EINECS: 206-017-1
Merck Index: 14,7960

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR