Sebacic acid 98+%
Supplier: Thermo Scientific Chemicals
A14158-0B
A14158-0I
A14158-22
A14158-30
CAAAAA14158-0BEA
152.47
CAD
CAAAAA14158-0B
CAAAAA14158-0I
CAAAAA14158-22
CAAAAA14158-30
Sebacic acid 98+%
Sebacic acid
Purity: 98+%
CAS No.: 111-20-6
Molecular Formula: C10H18O4
Formula Weight: 202.25
Flash Point: 220°(428°F)
Melting Point: 131-134°
Boiling Point: 295°/100mm
Beilstein Reference: 1210591
Merck Reference: 14,8415
MDL No.: MFCD00004440
Formula:
C₁₀H₁₈O₄ Boiling Pt: 295 °C/100 mm Melting Pt: 131…134 °C Density: 1.271 Flash Pt: 220 °C(428 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004440 CAS Number: 111-20-6 EINECS: 203-845-5 REACH: 01-2119519212-52 Merck Index: 14,8415 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...