1,10-Phenanthroline monohydrate 99+%

Supplier: Thermo Scientific Chemicals
Danger

A14140-09 A14140-18 A14140-30
CAAAAA14140-09EA 134.46 CAD
CAAAAA14140-09 CAAAAA14140-18 CAAAAA14140-30
1,10-Phenanthroline monohydrate 99+%
1,10-Phenanthroline monohydrate

Purity: 99+% CAS No.: 5144-89-8 Molecular Formula: C12H8N2.H2O Formula Weight: 198.23 (180.21anhy) Odor: Odorless Melting Point: 97-101° Beilstein Reference: 4008096 Merck Reference: 14,7214 UN No.: 2811 MDL No.: MFCD00149973

Formula: C₁₂H₈N₂·1H₂O
Melting Pt: 97…101 °C
Density: 1.259 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149973
CAS Number: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,7214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR