2-Chlorophenylhydrazinium chloride 97%

Supplier: Thermo Scientific Chemicals
Warning

A14136-06 A14136-14 A14136-22
CAAAA14136-06EA 47.87 CAD
CAAAA14136-06 CAAAA14136-14 CAAAA14136-22
2-Chlorophenylhydrazinium chloride 97%
2-Chlorophenylhydrazinium chloride

Purity: 97% CAS No.: 41052-75-9 Molecular Formula: C6H7ClN2.HCl Formula Weight: 179.05 Melting Point: ca 201° dec. Beilstein Reference: 3699381

Formula: C₆H₈Cl₂N₂
Melting Pt: ∼ 201 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012928
CAS Number: 41052-75-9
EINECS: 255-194-1
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR