2,4-Difluorobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

A14113-06 A14113-14
CAAAAA14113-06EA 55.77 CAD
CAAAAA14113-06 CAAAAA14113-14 CAAAAA14113-22
2,4-Difluorobenzonitrile 98%
2,4-Difluorobenzonitrile

Purity: 98% CAS No.: 3939-09-1 Molecular Formula: C7H3F2N Formula Weight: 139.11 Flash Point: 110°(230°F) Odor: Almond-like Boiling Point: 189° Beilstein Reference: 1940316 UN No.: 3439 MDL No.: MFCD00009826

Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 189 °C
Melting Pt: 43…48 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009826
CAS Number: 3939-09-1
EINECS: 223-523-8
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR