Bismuth sodium trioxide ≥80%

Supplier: Thermo Scientific Chemicals
Warning

A14073-14 A14073-22 A14073-36
CAAAAA14073-14EA 62.88 CAD
CAAAAA14073-14 CAAAAA14073-22 CAAAAA14073-36
Bismuth sodium trioxide ≥80%
Bismuth sodium trioxide

Purity: 80% CAS No.: 12232-99-4 Molecular Formula: NaBiO3 Formula Weight: 279.97 Odor: Odorless Merck Reference: 14,8585 MDL No.: MFCD00003474

Formula: BiNaO₃
Storage Temperature: Ambient
MDL Number: MFCD00003474
CAS Number: 12232-99-4
EINECS: 235-455-6
Merck Index: 14,8585

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR