Antimony(III) fluoride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Antimony trifluoride

A14068-0B A14068-18 A14068-30
CAAAAA14068-0BEA 502.44 CAD
CAAAAA14068-0B CAAAAA14068-18 CAAAAA14068-30
Antimony(III) fluoride 98%
Antimony (III) fluoride

Purity: 98% CAS No.: 7783-56-4 Molecular Formula: SbF3 Formula Weight: 178.75 Odor: Odorless Melting Point: 292° Boiling Point: 376° Merck Reference: 14,709 UN No.: 2923 MDL No.: MFCD00011218

Formula: F₃Sb
MW: 178.76 g/mol
Boiling Pt: 376 °C
Melting Pt: 292 °C
Density: 4.379
Storage Temperature: Ambient
MDL Number: MFCD00011218
CAS Number: 7783-56-4
EINECS: 232-009-2
UN: 2923
ADR: 8 III
Merck Index: 14,709

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR