1,5-Diphenylcarbazide 97+%

Supplier: Thermo Scientific Chemicals
Warning

A14060-14 A14060-22 A14060-36
CAAAAA14060-14EA 100.52 CAD
CAAAAA14060-14 CAAAAA14060-22 CAAAAA14060-36
1,5-Diphenylcarbazide 97+%
1,5-Diphenylcarbazide

Purity: 97+% CAS No.: 140-22-7 Molecular Formula: C13H14N4O Formula Weight: 242.28 Melting Point: 170-176° Beilstein Reference: 752039 Merck Reference: 14,3321 MDL No.: MFCD00003013

Formula: C₁₃H₁₄N₄O
Boiling Pt: 454 °C (1013 hPa)
Melting Pt: 170…176 °C
Density: 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003013
CAS Number: 140-22-7
EINECS: 205-403-7
Merck Index: 14,3321
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03cf8836a36f1077443ec2e3cfa151621925bd46&vid=a1b2bd876c3fbd9d6ded95c8430e4ec16bf5172b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR