m-Toluidine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14058-0B
A14058-30
CAAAAA14058-0BEA
165.9
CAD
CAAAAA14058-0B
CAAAAA14058-0I
CAAAAA14058-30
m-Toluidine 99%
m-Toluidine
Purity: 99%
CAS No.: 108-44-1
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 85°(185°F)
Refractive Index: 1.5670
Odor: Unpleasant
Melting Point: -30°
Boiling Point: 203-204°
Beilstein Reference: 635944
Merck Reference: 14,9536
UN No.: 1708
MDL No.: MFCD00007808
Formula:
C₇H₉N Boiling Pt: 203…204 °C Melting Pt: –30 °C Density: 0.992 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007808 CAS Number: 108-44-1 EINECS: 203-583-1 UN: 1708 ADR: 6.1,II Merck Index: 14,9536 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...