2-Naphthoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14046-09
A14046-18
A14046-30
CAAAA14046-09EA
101.44
CAD
CAAAA14046-09
CAAAA14046-18
CAAAA14046-30
2-Naphthoyl chloride 98%
2-Naphthoyl chloride
Purity: 98%
CAS No.: 2243-83-6
Molecular Formula: C11H7ClO
Formula Weight: 190.63
Flash Point:
Formula:
C₁₁H₇ClO Boiling Pt: 159…160 °C/9 mm Melting Pt: 48…52 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004093 CAS Number: 2243-83-6 EINECS: 218-822-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...