(Benzyl)triphenylphosphonium chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

A14034-22 A14034-36
CAAAAA14034-22EA 133.79 CAD
CAAAAA14034-22 CAAAAA14034-36
(Benzyl)triphenylphosphonium chloride 99%
(Benzyl)triphenylphosphonium chloride

Purity: 99% CAS No.: 1100-88-5 Molecular Formula: C6H5CH2P(C6H5)3Cl Formula Weight: 388.88 Flash Point: 300°(572°F) Odor: Odorless Beilstein Reference: 3599868 UN No.: 3464 MDL No.: MFCD00011913

Formula: C₂₅H₂₂ClP
Melting Pt: ∼ 330 °C (dec.)
Density: 1.18 g/cm³ (20 °C)
Flash Pt: >300 °C (572 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011913
CAS Number: 1100-88-5
EINECS: 214-154-3
UN: 3464
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR