D-(-)-Mannitol ≥97.0%, white powder

Supplier: Thermo Scientific Chemicals

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

A14030-0B A14030-0I A14030-30
CAAAAA14030-0BEA 114.39 CAD
CAAAAA14030-0B CAAAAA14030-0I CAAAAA14030-30
D-(-)-Mannitol ≥97.0%, white powder
D(-)-Mannitol

Purity: 99% CAS No.: 69-65-8 Molecular Formula: C6H14O6 Formula Weight: 182.17 Odor: Odorless Melting Point: 165-167° Boiling Point: 290-295°/3.5mm Beilstein Reference: 1721898 Merck Reference: 14,5745 Optical Rotation: +24° (in borax solution) MDL No.: MFCD00064287

Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Boiling Pt: 290…295 °C (3.5 mmHg)
Melting Pt: 165…167 °C
Density: 1.489
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
EINECS: 200-711-8
Merck Index: 14,5745
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR