Copper(II) nitrate hemipentahydrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14004-0B
A14004-0I
A14004-30
CAAAAA14004-0BEA
189.6
CAD
CAAAAA14004-0B
CAAAAA14004-0I
CAAAAA14004-30
Copper(II) nitrate hemipentahydrate 98%
Copper (II) nitrate hemipentahydrate
Purity: 98%
CAS No.: 19004-19-4
Molecular Formula: CuN2O6.2.5H2O
Formula Weight: 232.59 (187.55anhy)
Odor: Odorless
Melting Point: ca 115°
Boiling Point: 170° dec.
Merck Reference: 14,2643
UN No.: 3085
MDL No.: MFCD00149671
Formula:
Cu(NO₃)₂·2.5H₂O Boiling Pt: 170 °C (dec.) Melting Pt: ∼ 115 °C Density: 2.32 Storage Temperature: Ambient |
MDL Number:
MFCD02253001 CAS Number: 19004-19-4 EINECS: 221-838-5 UN: 3085 ADR: 5.1,III Merck Index: 14,2643 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...