3-Methyl-2-cyclopenten-1-one 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Methylcyclopent-2-enone, 3-Methyl-2-cyclopentenone
Stabilization:
Stabilised with hydroquinone (HQ) 0.1%
A13993-09
A13993-18
CAAAAA13993-09EA
140.15
CAD
CAAAAA13993-09
CAAAAA13993-18
3-Methyl-2-cyclopenten-1-one 98% stabilized
3-Methyl-2-cyclopenten-1-one
Purity: 98%
CAS No.: 2758-18-1
Molecular Formula: C6H8O
Formula Weight: 96.13
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Refractive Index: 1.4890
Odor: Pleasant
pH: 4.9
Melting Point: 2°
Boiling Point: 72-74°/15mm
Beilstein Reference: 1280476
MDL No.: MFCD00001403
Formula:
C₆H₈O MW: 96.13 g/mol Boiling Pt: 72…74 °C/15 mm Melting Pt: 2 °C Density: 0.980 Flash Pt: 65 °C(149 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001403 CAS Number: 2758-18-1 EINECS: 220-421-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...