Copper(II) sulfate, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Warning

A13986-0B A13986-0I A13986-30
CAAAAA13986-0BEA 404.48 CAD
CAAAAA13986-0B CAAAAA13986-0I CAAAAA13986-30
Copper(II) sulfate, anhydrous 98%
Copper (II) sulphate

Purity: 98% CAS No.: 7758-98-7 Molecular Formula: CuO4S Formula Weight: 159.60 Odor: Odorless Melting Point: 560° UN No.: 3288 MDL No.: MFCD00010981

Formula: CuSO₄
Melting Pt: >560 °C
Density: 3.6
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3077
ADR: 9,III
Merck Index: 14,2653

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR