Potassium-tert-butoxide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13947-0B
A13947-14
A13947-22
A13947-30
A13947-36
CAAAAA13947-0BEA
353.92
CAD
CAAAAA13947-0B
CAAAAA13947-14
CAAAAA13947-22
CAAAAA13947-30
CAAAAA13947-36
Potassium-tert-butoxide 97%
Potassium-tert-butoxide
Purity: 97%
CAS No.: 865-47-4
Molecular Formula: C4H9KO
Formula Weight: 112.22
Melting Point: ca 255° dec.
Beilstein Reference: 3556712
UN No.: 3206
MDL No.: MFCD00012162
Formula:
C₄H₉KO Melting Pt: ∼ 255 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012162 CAS Number: 865-47-4 EINECS: 212-740-3 UN: 3206 ADR: 4.2,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...