3,4-Dichlorophenylhydrazinium chloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4-Dichlorophenylhydrazine hydrochloride, 1-(3,4-Dichlorophenyl)hydrazine hydrochloride

A13906-09 A13906-18 A13906-30
CAAAA13906-09EA 120.24 CAD
CAAAA13906-09 CAAAA13906-18 CAAAA13906-30
3,4-Dichlorophenylhydrazinium chloride 98+%
3,4-Dichlorophenylhydrazinium chloride

Purity: 98+% CAS No.: 19763-90-7 Molecular Formula: C6H6Cl2N2.HCl Formula Weight: 213.50 Melting Point: ca 229° dec. Beilstein Reference: 3703934

Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
Melting Pt: ca 229 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012937
CAS Number: 19763-90-7
EINECS: 243-278-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR