Decahydronaphthalene (mixture of cis and trans isomers) 98%

Supplier: Thermo Scientific Chemicals
Danger

A13883-0D A13883-0F A13883-AE A13883-AP
CAAAAA13883-0DEA 987.5 CAD
CAAAAA13883-0D CAAAAA13883-0F CAAAAA13883-AE CAAAAA13883-AP
Decahydronaphthalene (mixture of cis and trans isomers) 98%
Decahydronaphthalene (mixture of cis and trans isomers)

Purity: 98% CAS No.: 91-17-8 Molecular Formula: C10H18 Formula Weight: 138.25 Flash Point: 57°(134°F) Refractive Index: 1.4740 Odor: Mild Melting Point: -45° Boiling Point: 189-191° Beilstein Reference: 878165 Merck Reference: 14,2846 UN No.: 1147 MDL No.: MFCD00004130

Formula: C₁₀H₁₈
Boiling Pt: 189…191 °C
Melting Pt: < –45 °C
Density: 0.88
Flash Pt: 57 °C(134 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004130
CAS Number: 91-17-8
EINECS: 202-046-9
UN: 1147
ADR: 3,III
Merck Index: 14,2846
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41b5fff8aa81be0e42af177a8f0e3ac149943c6e&vid=ca9f3c5d71ac621a6216f8b8b28d7cb27b29fd84&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR