Oxalic acid dihydrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13866-0B
A13866-0I
A13866-30
CAAAAA13866-0BEA
162.74
CAD
CAAAAA13866-0B
CAAAAA13866-0I
CAAAAA13866-30
Oxalic acid dihydrate 98%
Oxalic acid dihydrate
Purity: 98%
CAS No.: 6153-56-6
Molecular Formula: C2H2O4.2H2O
Formula Weight: 126.07 (90.04anhy)
Odor: Odorless
pH: 1.3
Melting Point: 101-105°
Beilstein Reference: 3679436
Merck Reference: 14,6911
UN No.: 3261
MDL No.: MFCD00149102
Formula:
HO₂CCO₂H·2H₂O Boiling Pt: 149 - 160 °C (1013 hPa) Melting Pt: 101…105 °C Density: 1.65 Storage Temperature: Ambient |
MDL Number:
MFCD00149102 CAS Number: 6153-56-6 EINECS: 205-634-3 ADR: 8,III REACH: 01-2119534576-33 Merck Index: 14,6911 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...