Diethyl ethoxymethylenemalonate 98+%

Supplier: Thermo Scientific Chemicals
Danger

A13776-22 A13776-36 A13776-18
CAAAAA13776-22EA 55.62 CAD
CAAAAA13776-22 CAAAAA13776-36 CAAAA13776-18 CAAAA13776-30
Diethyl ethoxymethylenemalonate 98+%
Diethyl ethoxymethylenemalonate

Purity: 98+% CAS No.: 87-13-8 Molecular Formula: C10H16O5 Formula Weight: 216.23 Flash Point: 144°(291°F) Refractive Index: 1.4620 Odor: Ester-like Melting Point: -33° Boiling Point: 136-139°/10mm Beilstein Reference: 880058 MDL No.: MFCD00009148

Formula: C₁₀H₁₆O₅
Boiling Pt: 136…139 °C/10 mm
Melting Pt: –33 °C
Density: 1,080
Flash Pt: 144 °C(291 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009148
CAS Number: 87-13-8
EINECS: 201-725-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR