Geraniol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13736-0B
A13736-18
A13736-30
CAAAAA13736-0BEA
737.86
CAD
CAAAAA13736-0B
CAAAAA13736-18
CAAAAA13736-30
Geraniol 97%
Geraniol
Purity: 97%
CAS No.: 106-24-1
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 101°(213°F)
Refractive Index: 1.4760
Odor: Floral
Melting Point: -15°
Boiling Point: 114-115°/12mm
Beilstein Reference: 1722456
Merck Reference: 14,4403
MDL No.: MFCD00002917
Formula:
C₁₀H₁₈O Boiling Pt: 114…115 °C/12 mm Melting Pt: –15 °C Density: 0.884 Flash Pt: 101 °C(213 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002917 CAS Number: 106-24-1 EINECS: 203-377-1 Merck Index: 14,4403 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...