Potassium thiocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13731-0E
A13731-36
CAAAAA13731-0EEA
437.66
CAD
CAAAAA13731-0E
CAAAAA13731-36
Potassium thiocyanate 98%
Potassium thiocyanate
Purity: 98%
CAS No.: 333-20-0
Molecular Formula: KSCN
Formula Weight: 97.18
Odor: Odorless
pH: 5.3-8.5
Melting Point: 172-175°
Boiling Point: 500° dec.
Beilstein Reference: 3594799
Merck Reference: 14,7691
MDL No.: MFCD00011413
Formula:
KSCN Boiling Pt: 500 °C (dec.) Melting Pt: 172…175 °C Density: 1.89 Storage Temperature: Ambient |
MDL Number:
MFCD00011413 CAS Number: 333-20-0 EINECS: 206-370-1 Merck Index: 14,7691 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...