5-Nonanone 98%
Supplier: Thermo Scientific Chemicals
Warning
|
A13722-22
A13722-36
CAAAAA13722-22EA
99.54
CAD
CAAAAA13722-22
CAAAAA13722-36
5-Nonanone 98%
5-Nonanone
Purity: 98%
CAS No.: 502-56-7
Molecular Formula: C9H18O
Formula Weight: 142.24
Flash Point: 65°(149°F)
Refractive Index: 1.4190
Melting Point: -50°
Boiling Point: 186-187°
Beilstein Reference: 1743583
MDL No.: MFCD00009484
Formula:
C₉H₁₈O Boiling Pt: 186…187 °C Melting Pt: –50 °C Density: 0.824 Flash Pt: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009484 CAS Number: 502-56-7 EINECS: 207-946-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...