2-Fluorophenylhydrazinium chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A13716-03 A13716-06
CAAAA13716-03EA 41.24 CAD
CAAAA13716-03 CAAAA13716-06 CAAAA13716-14
2-Fluorophenylhydrazinium chloride 98%
2-Fluorophenylhydrazinium chloride

Purity: 98% CAS No.: 2924-15-4 Molecular Formula: C6H7FN2.HCl Formula Weight: 162.60 Melting Point: ca 205° dec. Beilstein Reference: 3627985

Formula: C₆H₈ClFN₂
Melting Pt: ∼ 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012927
CAS Number: 2924-15-4
EINECS: 220-885-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR