Titanium(IV) isopropoxide 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13703-0E
A13703-22
A13703-36
CAAAAA13703-0EEA
415.54
CAD
CAAAAA13703-0E
CAAAAA13703-22
CAAAAA13703-36
Titanium(IV) isopropoxide 95%
Titanium (IV) isopropoxide
Purity: 95%
CAS No.: 546-68-9
Molecular Formula: C12H28O4Ti
Formula Weight: 284.23
Flash Point: 46°(115°F)
Refractive Index: 1.4640
Melting Point: 16-20°
Boiling Point: 232°
Beilstein Reference: 3679474
Merck Reference: 14,9480
UN No.: 2413
MDL No.: MFCD00008871
Formula:
C₁₂H₂₈O₄Ti Boiling Pt: 232 °C Melting Pt: 16…20 °C Density: 0.955 Flash Pt: 46 °C (115 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008871 CAS Number: 546-68-9 EINECS: 208-909-6 UN: 2413 ADR: 3,III Merck Index: 14,9480 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...