L(+)-Tartaric acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13668-0E
A13668-22
A13668-36
CAAAAA13668-0EEA
188.04
CAD
CAAAAA13668-0E
CAAAAA13668-22
CAAAAA13668-36
L(+)-Tartaric acid 99%
L(+)-Tartaric acid
Purity: 99%
CAS No.: 87-69-4
Molecular Formula: C4H6O6
Formula Weight: 150.09
Odor: Odorless
pH: 2.1
Melting Point: 168-170°
Beilstein Reference: 1725147
Merck Reference: 14,9070
Optical Rotation: +12.4° (c=20 in water)
MDL No.: MFCD00064207
Formula:
C₄H₆O₆ Boiling Pt: 399 °C (1013 hPa) Melting Pt: 168…170 °C Density: 1.760 Flash Pt: (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00064207 CAS Number: 87-69-4 EINECS: 201-766-0 REACH: 01-2119537204-47 Merck Index: 14,9070 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...