Chlorotrimethylsilane 98+% (by GC)
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13651-AC
A13651-AE
A13651-AP
CAAAAA13651-ACEA
41.24
CAD
CAAAAA13651-AC
CAAAAA13651-AE
CAAAAA13651-AP
Chlorotrimethylsilane 98+% (by GC)
Chlorotrimethylsilane
Purity: 98+%
CAS No.: 75-77-4
Molecular Formula: C3H9ClSi
Formula Weight: 108.64
Flash Point: -27°(-16°F)
Refractive Index: 1.3887
Odor: Chlorine-like
Melting Point: -58°
Boiling Point: 56-58°
Beilstein Reference: 1209232
UN No.: 1298
MDL No.: MFCD00000502
Formula:
C₃H₉ClSi Boiling Pt: 56…58 °C Melting Pt: –58 °C Density: 0.856 Flash Pt: –27 °C (–16 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000502 CAS Number: 75-77-4 EINECS: 200-900-5 UN: 1298 ADR: 3,II REACH: 01-2119457596-25 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...