2-Ethoxyethyl acetate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethylglycol acetate
A13639-0F
A13639-AP
CAAAAA13639-0FEA
122.29
CAD
CAAAAA13639-0F
CAAAAA13639-AP
2-Ethoxyethyl acetate 98+%
2-Ethoxyethyl acetate
Purity: 98+%
CAS No.: 111-15-9
Molecular Formula: C6H12O3
Formula Weight: 132.16
Flash Point: 52°(126°F)
Refractive Index: 1.4070
Odor: Fruit-like
Melting Point: -62°
Boiling Point: 155-157°
Beilstein Reference: 1748677
Merck Reference: 14,3751
UN No.: 1172
MDL No.: MFCD00009251
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol Boiling Pt: 155…157 °C Melting Pt: –62 °C Density: 0.975 Flash Pt: 52 °C (126 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009251 CAS Number: 111-15-9 EINECS: 203-839-2 UN: 1172 ADR: 3,III Merck Index: 14,3751 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...