2-(4-Chlorophenyl)indole 98%

Supplier: Thermo Scientific Chemicals

A13638-03 A13638-06
CAAAA13638-03EA 54.04 CAD
CAAAA13638-03 CAAAA13638-06
2-(4-Chlorophenyl)indole 98%
2-(4-Chlorophenyl)indole

Purity: 98% CAS No.: 1211-35-4 Molecular Formula: C14H10ClN Formula Weight: 227.70 Melting Point: 205-207° MDL No.: MFCD00047159

Formula: C₁₄H₁₀ClN
Melting Pt: 205…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00047159
CAS Number: 1211-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR