2-(4-Chloro-3-nitrobenzoyl)benzoic acid 98%
Supplier: Thermo Scientific Chemicals
A13605-18
A13605-30
CAAAA13605-18EA
103.17
CAD
CAAAA13605-18
CAAAA13605-30
2-(4-Chloro-3-nitrobenzoyl)benzoic acid 98%
2-(4-Chloro-3-nitrobenzoyl)benzoic acid
Purity: 98%
CAS No.: 85-54-1
Molecular Formula: C14H8ClNO5
Formula Weight: 305.68
Melting Point: 198-201°
MDL No.: MFCD00007082
Formula:
C₁₄H₈ClNO₅ MW: 305.67 g/mol Melting Pt: 198…201 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007082 CAS Number: 85-54-1 EINECS: 201-613-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...