Methanesulfonic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13565-0E
A13565-22
A13565-36
CAAAAA13565-0EEA
426.6
CAD
CAAAAA13565-0E
CAAAAA13565-22
CAAAAA13565-36
Methanesulfonic acid 98+%
Methanesulphonic acid
Purity: 98+%
CAS No.: 75-75-2
Molecular Formula: CH4O3S
Formula Weight: 96.10
Flash Point: 156°(312°F)
Refractive Index: 1.4300
Melting Point: 19-20°
Boiling Point: 166-168°/10mm
Beilstein Reference: 1446024
Merck Reference: 14,5954
UN No.: 2586
MDL No.: MFCD00007518
Formula:
CH₃SO₃H MW: 96.11 g/mol Boiling Pt: 166…168 °C/10 mm Melting Pt: 19…20 °C Density: 1.481 Flash Pt: 156 °C (312 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007518 CAS Number: 75-75-2 EINECS: 200-898-6 UN: 3265 ADR: 8,III Merck Index: 14,5954 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...