4-(4-Nitrobenzyl)pyridine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(p-Nitrobenzyl)Pyridine

A13564-06 A13564-14 A13564-22
CAAAA13564-06EA 70.63 CAD
CAAAA13564-06 CAAAA13564-14 CAAAA13564-22
4-(4-Nitrobenzyl)pyridine 98+%
4-(4-Nitrobenzyl)pyridine

Purity: 98+% CAS No.: 1083-48-3 Molecular Formula: C12H10N2O2 Formula Weight: 214.22 Melting Point: 69-72° Beilstein Reference: 170877

Formula: C₁₂H₁₀N₂O₂
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00006445
CAS Number: 1083-48-3
EINECS: 214-108-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR