N,N-Dimethylformamide 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF
A13547-0D
A13547-0F
A13547-AK
CAAAAA13547-0DEA
470.84
CAD
CAAAAA13547-0D
CAAAAA13547-0F
CAAAAA13547-AK
N,N-Dimethylformamide 99%
N,N-Dimethylformamide
Purity: 99%
CAS No.: 68-12-2
Molecular Formula: HCON(CH3)2
Formula Weight: 73.09
Flash Point: 57°(135°F)
Refractive Index: 1.4310
Odor: Amine-like
pH: 7
Melting Point: -61°
Boiling Point: 153°
Beilstein Reference: 605365
Merck Reference: 14,3243
UN No.: 2265
MDL No.: MFCD00003284
Formula:
HCON(CH₃)₂ MW: 73.09 g/mol Boiling Pt: 153 °C Melting Pt: –61 °C Density: 0.944 Flash Pt: 57 °C (135 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3,III REACH: 01-2119475605-32 Merck Index: 14,3243 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...