Guanidinium hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

A13543-0B A13543-0I A13543-30
CAAAAA13543-0BEA 157.53 CAD
CAAAAA13543-0B CAAAAA13543-0I CAAAAA13543-30
Guanidinium hydrochloride 98%
Guanidine hydrochloride

Purity: 98% CAS No.: 50-01-1 Molecular Formula: CH5N3.HCl Formula Weight: 95.53 Melting Point: 178-185° Beilstein Reference: 3591990 Merck Reference: 14,4562 MDL No.: MFCD00013026

Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 178…185 °C
Density: 1.344
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 14,4562
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR