p-Toluenesulfonyl hydrazide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methylbenzenesulfonohydrazide, 4-Methylbenzenesulphonylhydrazide, 4-Toluenesulphonyl hydrazide, 4-Methylbenzenesulphonhydrazide, Toluene-4-sulfonohydrazide, 4-Methylbenzenesulphonic acid hydrazide, 4-Toluenesulfonyl hydrazide, 4-Methylbenzenesulphonohydrazide, Toluene-4-sulphonohydrazide, 4-Methylbenzenesulfonic acid hydrazide

A13529-14 A13529-22 A13529-36
CAAAAA13529-14EA 50.4 CAD
CAAAAA13529-14 CAAAAA13529-22 CAAAAA13529-36
p-Toluenesulfonyl hydrazide 98%
p-Toluenesulphonyl hydrazide

Purity: 98% CAS No.: 1576-35-8 Molecular Formula: C7H10N2O2S Formula Weight: 186.23 Melting Point: 108-111° Beilstein Reference: 610130 UN No.: 3226 MDL No.: MFCD00007588

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 108…111 °C
Density: 1.4
Storage Temperature: Ambient
MDL Number: MFCD00007588
CAS Number: 1576-35-8
EINECS: 216-407-3
UN: 3226
ADR: 4.1,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR