2,5-Dichlorobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Danger

A13525-06 A13525-14
CAAAA13525-06EA 156.1 CAD
CAAAA13525-06 CAAAA13525-14
2,5-Dichlorobenzaldehyde 98%
2,5-Dichlorobenzaldehyde

Purity: 98% CAS No.: 6361-23-5 Molecular Formula: C7H4Cl2O Formula Weight: 175.01 Flash Point: 110°(230°F) Melting Point: 53-58° Boiling Point: 231-233° Beilstein Reference: 1635476 UN No.: 1759 MDL No.: MFCD00156140

Formula: C₇H₄Cl₂O
Boiling Pt: 231…233 °C
Melting Pt: 53…58 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00156140
CAS Number: 6361-23-5
EINECS: 228-832-1
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR