trans-Anethole 98+% (by GC) stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(E)-1-Methoxy-4-(prop-1-en-1-yl)benzene
A13482-14
A13482-22
A13482-36
CAAAAA13482-14EA
60.2
CAD
CAAAAA13482-14
CAAAAA13482-22
CAAAAA13482-36
trans-Anethole 98+% (by GC) stabilized
trans-Anethole
Inhibits lung and forestomach carcinogenesis
Purity: 98+%
CAS No.: 4180-23-8
Molecular Formula: C10H12O
Formula Weight: 148.21
Storage Temperature: 2°C to 8°C
Flash Point: 96°(205°F)
Refractive Index: 1.5610
Melting Point: 20-23°
Boiling Point: 232-235°
Beilstein Reference: 774229
Merck Reference: 14,643
MDL No.: MFCD00009284
Formula:
C₁₀H₁₂O MW: 148.2 g/mol Boiling Pt: 232…235 °C Melting Pt: 20…23 °C Density: 0.991 Flash Pt: 96 °C (205 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009284 CAS Number: 4180-23-8 EINECS: 224-052-0 Merck Index: 14,643 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...