Ethyl-4-bromobutyrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4-bromobutyrate

A13470-0B A13470-18 A13470-30
CAAAA13470-0BEA 805.8 CAD
CAAAA13470-0B CAAAA13470-18 CAAAA13470-30
Ethyl-4-bromobutyrate 98%
Ethyl 4-bromobutyrate

Purity: 98% CAS No.: 2969-81-5 Molecular Formula: Br(CH2)3CO2CH2CH3 Formula Weight: 195.06 Flash Point: 90°(194°F) Physical Form: Liquid Density: 1.363 Refractive Index: 1.4560 Appearance: Colorless to pale yellow Odor: Odorless Boiling Point: 80-82°/10mm Beilstein Reference: 1749700 MDL No.: MFCD00000259

Formula: C₆H₁₁BrO₂
MW: 195.06 g/mol
Boiling Pt: 80…82 °C/10 mm
Density: 1,363
Flash Pt: 90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000259
CAS Number: 2969-81-5
EINECS: 221-005-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR