Sulfolane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetramethylene sulfone, Tetrahydrothiophene-1,1-dioxide, Tetramethylene sulphone, Tetrahydrothiophene-1,1-dioxide, Tetrahydrothiophene 1,1-dioxide
A13466-0B
A13466-0I
A13466-30
CAAAA13466-0BEA
165.9
CAD
CAAAA13466-0B
CAAAA13466-0I
CAAAA13466-30
Sulfolane 99%
Sulpholane
Purity: 99%
CAS No.: 126-33-0
Molecular Formula: C4H8O2S
Formula Weight: 120.17
Flash Point: 166°(330°F)
Refractive Index: 1.4825
Odor: Weak, characteristic
Melting Point: 25-28°
Boiling Point: 285°
Beilstein Reference: 107765
Merck Reference: 14,8955
MDL No.: MFCD00005484
Formula:
C₄H₈O₂S MW: 120.17 g/mol Boiling Pt: 285 °C Melting Pt: 25…28 °C Density: 1.261 Flash Pt: 166 °C(330 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005484 CAS Number: 126-33-0 EINECS: 204-783-1 UN: 3335 ADR: "9", Merck Index: 14,8955 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...