(E)-β-Ionone 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
(E)-4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one, (E)-4-(2,6,6-Trimethylcyclohex-1-en-1-yl)but-3-en-2-one, beta-Ionone
A13400-22
A13400-36
CAAAA13400-22EA
93.06
CAD
CAAAA13400-22
CAAAA13400-36
(E)-β-Ionone 96%
β-Ionone
Purity: 96%
CAS No.: 79-77-6
Molecular Formula: C13H20O
Formula Weight: 192.30
Flash Point: 112°(233°F)
Refractive Index: 1.5200
Melting Point: -49°
Boiling Point: 126-128°/12mm
Beilstein Reference: 1909544
Merck Reference: 14,5056
MDL No.: MFCD00001549
Formula:
C₁₃H₂₀O MW: 192.3 Boiling Pt: 126…128 °C (12 mmHg) Melting Pt: –49 °C Density: 0.943 Flash Pt: 112 °C (233 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001549 CAS Number: 79-77-6 EINECS: 201-224-3 Merck Index: 14,5056 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...