1-Bromo-3-methyl-2-butene 90+% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with silver
A13388-09
A13388-18
A13388-30
CAAAA13388-09EA
71.1
CAD
CAAAA13388-09
CAAAA13388-18
CAAAA13388-30
1-Bromo-3-methyl-2-butene 90+% stabilized
1-Bromo-3-methyl-2-butene
Purity: 90+%
CAS No.: 870-63-3
Molecular Formula: C5H9Br
Formula Weight: 149.03
Storage Temperature: 2°C to 8°C
Flash Point: 32°(90°F)
Refractive Index: 1.4895
Odor: Pungent
Boiling Point: 135°
Beilstein Reference: 1420778
UN No.: 2920
MDL No.: MFCD00000242
Formula:
C₅H₉Br Boiling Pt: 135 °C Density: 1.281 Flash Pt: 32 °C (90 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000242 CAS Number: 870-63-3 EINECS: 212-799-5 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...