6-Methyl-2,4-heptanedione 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13373-04
A13373-09
A13373-18
CAAAAA13373-04EA
50.56
CAD
CAAAAA13373-04
CAAAAA13373-09
CAAAAA13373-18
6-Methyl-2,4-heptanedione 98+%
6-Methyl-2,4-heptanedione
Purity: 98+%
CAS No.: 3002-23-1
Molecular Formula: C8H14O2
Formula Weight: 142.20
Flash Point: 58°(136°F)
Refractive Index: 1.4560
Boiling Point: 183°
Beilstein Reference: 1751122
UN No.: 1224
MDL No.: MFCD00008939
Formula:
C₈H₁₄O₂ Boiling Pt: 183 °C Density: 0.919 Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008939 CAS Number: 3002-23-1 EINECS: 221-093-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...