Diphosphorus pentoxide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phosphorus(V) oxide, di-Phosphorus pentoxide, Phosphorus pentoxide, Phosphoric anhydride

A13348-0E A13348-36
CAAAAA13348-0EEA 175.38 CAD
CAAAAA13348-0E CAAAAA13348-36
Diphosphorus pentoxide 98%
di-Phosphorus pentoxide

Purity: 98% CAS No.: 1314-56-3 Molecular Formula: P2O5 Formula Weight: 141.94 Odor: Weak Melting Point: 340° Boiling Point: 360° subl. Merck Reference: 14,7355 UN No.: 1807 MDL No.: MFCD00011440

Formula: P₂O₅
MW: 141.94 g/mol
Boiling Pt: 360 °C (subl.)
Melting Pt: 340 °C
Density: 2.39
Storage Temperature: Ambient
MDL Number: MFCD00011440
CAS Number: 1314-56-3
EINECS: 215-236-1
UN: 1807
ADR: 8,II
Merck Index: 14,7355

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR