Pentafluorophenylhydrazine 97%

Supplier: Thermo Scientific Chemicals

A13302-06 A13302-14
CAAAA13302-06EA 70.15 CAD
CAAAA13302-06 CAAAA13302-14
Pentafluorophenylhydrazine 97%
Pentafluorophenylhydrazine

Purity: 97% CAS No.: 828-73-9 Molecular Formula: C6H3F5N2 Formula Weight: 198.09 Melting Point: 73-78° Beilstein Reference: 747800

Formula: C₆H₃F₅N₂
Melting Pt: 73…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00007574
CAS Number: 828-73-9
EINECS: 212-586-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR